Would you like to log in again?
Durx® 670 Cellulose/Polyester Nonwoven Disc Wipes, Berkshire
Supplier: Berkshire
Durx®
DR670.03D.32
76359-588CS 531.46 USD InStock
76359-588
Durx® 670 Cellulose/Polyester Nonwoven Disc Wipes, Berkshire
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Durx® 670 disc wipes provide an excellent combination of the synthetic polyester strength and cleanliness with the absorbent characteristics of cellulose.
  • Hydroentangled 55% cellulose/45% polyester
  • Smooth, highly absorbent, and durable
  • Low extractable levels and particle counts
  • Knife-cut edges
  • Chemically compatible with common cleaning and disinfecting solutions
  • Double packed
  • Contains no latex
Superior strength and absorbency make Durx® 670 an ideal choice for a general-purpose disc wipe. Manufactured without the use of binders or surfactants, ion levels and organic extractables remain low. These can be used for cleaning and maintenance in laboratories, aerospace, defense electronics, food processing, computer assembly and semiconductor, circuit board, pharmaceutical, and biomedical device manufacture.
No secondary binders. Critical combination of the synthetic polyester strength with the absorbent characteristics of cellulose. Wipes work well with IPA and other solvents. The absorbent, low particle work surface material is ideal for cleaning work surfaces and spill pickup. Also, well suited for general-purpose wiping and polishing of surface areas, including optics. Steam autoclavable for use in aseptic areas.
Certifications: Designed for use in ISO class 5 and higher cleanroom environments. EC regulation 1935 (2004)-Food contact compliant. FDA 21 CFR-Food contact compliant. USP <797> applications.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty