Would you like to log in again?
Cryopres™ Cell Cryopreservation Medium, MP Biomedicals
Supplier: MP Biomedicals
092780248
76337-132EA 180.35 USD InStock
76337-132
Cryopres™ Cell Cryopreservation Medium, MP Biomedicals
Cell Culture Supplements and Additives
Cell cryopreservation medium is a balanced, protein free, ready-to-use cryopreservation solution with 10% USP DMSO (Cryopres™).
It does not require any dilution or further processing and can be used for cells cultured in media with or without serum.
Caution: For laboratory use. Not for injection.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty