Would you like to log in again?
Myco-Sniff™ Mycoplasma PCR Kit, MP Biomedicals
Supplier: MP Biomedicals
093050201
76337-168EA 375.44 USD InStock
76337-168
Myco-Sniff™ Mycoplasma PCR Kit, MP Biomedicals
Assays qPCR Assays
Myco-Sniff™ mycoplasma PCR detection kit provides a highly sensitive, specific and rapid method for the detection of a broad range of mycoplasma species in cell cultures.
  • Highly specific: No Interference of animal or bacterial DNA
  • Sample control: Easily verify the effectiveness of template gDNA by checking the amplification of the sample control DNA
The detection limit for contaminating mycoplasma is as low as 20 CFU/ml. Includes 48 lyophilized PCR tubes for 20 μl reactions.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty