Would you like to log in again?
InTemp® Bluetooth® Temperature Probe with Dataloggers
Supplier: ONSET COMPUTER CORPORATION SE
Bluetooth®
CX402-T2M CX402-T4M
470323-324EA 223.79 USD InStock
470323-324 470323-326
InTemp® Bluetooth® Temperature Probe with Dataloggers
Data Loggers
InTemp® CX402 data loggers monitor temperatures in refrigerators, freezers, and other temperature-controlled environments to comply with global GDP requirements.
  • Easy-to-read display with current and daily min/max temperatures
  • Bluetooth® transmission of data and alarm states
  • Logger setup and download of data in seconds – without a PC or cable
  • Real-time alerts and automatic data downloads with optional CX Gateway device
  • ±0.5 °C accuracy for reliable temperature readings
  • Battery-powered to guard against local power loss
  • NIST Certificate of Calibration
The loggers communicate wirelessly via Bluetooth® Low Energy to mobile devices. Using the InTemp app, you can easily view data, check logger status, set alarms, and create and share secure PDF reports for streamlined reporting and regulatory compliance. All loggers feature a built-in LCD display to view current and daily min and max temperatures, advanced audible and visual alarm capabilities for notification of temperature excursions, an internal sensor for ambient temperature monitoring, and a single-point NIST Certification of Calibration for both the logger body and probe.
Models are available with a two or four meter external temperature probe for tracking actual refrigerator temperatures without Glycol buffers or freezer applications.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty