Would you like to log in again?
InTemp® Bluetooth® Ambient Temperature Datalogger
Supplier: ONSET COMPUTER CORPORATION SE
Bluetooth®
CX403
470323-334EA 170.79 USD InStock
470323-334
InTemp® Bluetooth® Ambient Temperature Datalogger
Data Loggers
The InTemp® CX403 data logger features an internal sensor for monitoring ambient temperature.
  • Easy-to-read display with current and daily min/max temperatures
  • Bluetooth transmission of data and alarm states
  • Logger setup and download of data in seconds – without a PC or cable
  • Real-time alerts and automatic data downloads with optional CX Gateway device
  • ±0.5 °C accuracy for reliable temperature readings
  • Battery-powered to guard against local power loss
The logger communicates wirelessly via Bluetooth® Low Energy to mobile devices. The CX403 features a built-in LCD display to view ambient temperature, advanced audible and visual alarm capabilities for notification of temperature excursions, and a NIST Certification of Calibration.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty