Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 4 mol/l (4 N) in aqueous solution, Titripur® volumetric solution, Supelco®

Supplier: MilliporeSigma
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

M-Clarity™ quality level = MQ100

1.11584.5000 1.11584.5000
EM1.11584.5000CS 518 USD InStock
EM1.11584.5000 CA1.11584.5000
Sodium hydroxide 4 mol/l (4 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Sodium hydroxide


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 109 °C (1013 hPa)
Density: ≤1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,
Order Now
Specification Test Results
Form liquid
Amount-of-substance concentration 3,980-4,020
Measurement uncertainty + / - 0,012
Traceability NIST SRM
The concentration is determined by volumetric titration and refers to 20°C.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty