Would you like to log in again?
Diamond® RealSeal™ Bottles, Amber, Narrow Mouth, Polypropylene, Globe Scientific
Supplier: Globe Scientific
7058000AM 7052000AM 7054000AM
76360-352EA 38.93 USD InStock
76360-352 76360-354 76360-356
Diamond® RealSeal™ Bottles, Amber, Narrow Mouth, Polypropylene, Globe Scientific
Bottles Plastic Bottles
Diamond® RealSeal™ Plastic Laboratory Bottles from Globe Scientific meet the requirements of the most demanding applications. Rugged and durable, these bottles are a safe, shatterproof alternative to glass bottles for collection, storage, or shipping samples, media, and reagents.
  • Narrow mouth bottles are perfect for collecting and storing liquids
  • 4 and 8 L bottles have an integrated shoulder loop for attaching and ID tag
  • Suitable for autoclaving at 121 °C
  • Improved chemical resistance and clarity
  • Resistant to most corrosives and suitable for a wide variety of laboratory uses
  • Amber polypropylene bottles reduce UV Light transmission to protect light sensitive contents during storage and transport
  • Amber bottles comply with US Pharmacopeia requirements for maximum light transmission
Diamond® RealSeal™ Bottles are available in a wide range of shapes, sizes, and materials to accomodate most application needs. Small or large, wide mouth or narrow, natural or amber - even large format with handles - the Diamond® RealSeal™ line includes the most frequently used and less common bottles.
Diamond RealSeal™ Large Format Plastic Laboratory Bottles from Globe Scientific meet the requirements of the most demanding applications. These new additions expand the application range to volumes up to 8 liters with several material and design options.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty