Would you like to log in again?
Lumbar Puncture Trays, BD
Supplier: BD Medical
4303C 4301C 4302C
PH4303CCS 772.78 USD InStock
PH4303C PH4302C PH4301C
Lumbar Puncture Trays, BD
Trays/Dishes/Pots Instrument Trays/Dishes
These Lumbar trays contain components to measure cerebrospinal fluid (CSF) pressure and collect CSF specimens.
Lumbar trays available with atraumatic needles, which are recommended by the American Academy of Neurology to reduce post-lumbar puncture headaches.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty