Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Taxo™ Anaerobe Differentiation Disc Set, BD Diagnostics
Supplier: BD Diagnostics
231651
BB31651PK 168.47 USD InStock
BB31651
Taxo™ Anaerobe Differentiation Disc Set, BD Diagnostics
Microbiology Test Systems Antibiotic Tests
BD Taxo™ anaerobe differentiation disc set for the differentiation of Gram-negative bacilli.
Delivery information: Set includes one cartridge each of: Kanamycin one mg K; Rifampin 15 µg RA; Penicillin two units P-2; Vancomycin five µg Va-5, Colistin ten µg CL-10.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty