Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CALB1 Chicken Polyclonal Antibody

Supplier: Biosensis
C-1798-50
76333-952EA 341 USD InStock
76333-952
Anti-CALB1 Chicken Polyclonal Antibody
Antibodies
Buffers cytosolic calcium. May stimulate a membrane Ca2+-ATPase and a 3',5'-cyclic nucleotide phosphodiesterase. Ref: uniprot.org

Antibody is specific for calbindin and does not recognize closely related proteins parvalbumin and calretinin as determined by Western Blotting.

Applications: Western blotting (1:1,000-1:5,000) and Immunohistochemistry (1:1,000-1:5,000). Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CALB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, cow, rat, mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty