Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NEST Chicken Polyclonal Antibody

Supplier: Biosensis
C-1812-50
76333-980EA 341 USD InStock
76333-980
Anti-NEST Chicken Polyclonal Antibody
Antibodies
Required for brain and eye development. Promotes the disassembly of phosphorylated vimentin intermediate filaments (IF) during mitosis and may play a role in the trafficking and distribution of IF proteins and other cellular factors to daughter cells during progenitor cell division. Required for survival, renewal and mitogen-stimulated proliferation of neural progenitor cells (By similarity). Ref: uniprot.org

Applications: Western blotting (1:1,000-1:5,000) and Immunocytochemistry (1:2,000-1:5,000). Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: NEST
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino acids 315-630
Host: Chicken
Isotype: IgY
Reactivity: Human, rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty