Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CN37 Mouse Monoclonal Antibody [clone: 1H10]

Supplier: Biosensis
M-1802-50
76334-016EA 341 USD InStock
76334-016
Anti-CN37 Mouse Monoclonal Antibody [clone: 1H10]
Antibodies
May participate in RNA metabolism in the myelinating cell, CNP is the third most abundant protein in central nervous system myelin. Ref: uniprot.org

Applications: Western blotting (1:5,000-1:20,000), Immunocytochemistry (1:1,000-1:5,000) and Immunohistochemistry (1:500-1:5,000). Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CN37
Clonality: Monoclonal
Clone: 1H10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, rat, mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty