Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

KIMBLE® 7/12 Standard Taper KONTES® Thermometer, DWK Life Sciences

Supplier: DWK Life Sciences
KIMBLE®
308900-0211
KT308900-0211EA 65.37 USD InStock
KT308900-0211
KIMBLE® 7/12 Standard Taper KONTES® Thermometer, DWK Life Sciences
Thermometers Analog Thermometers
Laboratory-grade thermometer with proportions smaller than standard KT871000-0925 standard taper thermometers. Therefore, less heat transfer occurs from the sample to the thermometer.

  • White back
  • Subdivided in 1 °C increments
  • Accurate to ±1 °C
  • Contains 3 g Hg

Standard taper 7/12 joint will assemble with a 10/18 standard taper joint when a adapter is used.

Caution: This item contains mercury. Exposed mercury can evaporate and become an invisible, odorless toxic vapor. Use caution in storing and handling this product. Products containing mercury should not be put in the trash; they must be recycled or disposed of as hazardous waste according to state and local laws.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty