Would you like to log in again?
BBL™ Corn Meal Agar Deeps with Polysorbate 80 Pour Tube, BD
Supplier: BD
297235
90004-784PK 25.99 USD InStock
90004-784
BBL™ Corn Meal Agar Deeps with Polysorbate 80 Pour Tube, BD
Media for Microbiology Prepared Media for Microbiology
Corn Meal Agar promotes the development of various microscopic morphological structures by yeasts that aid in their identification.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty