Would you like to log in again?
Industrial Wipes, Non-Woven, FG Cleanwipes
Supplier: FG Cleanwipes
799CBDB00
76244-914CS 461 USD InStock
76244-914
Industrial Wipes, Non-Woven, FG Cleanwipes
Wipes Disposable Wipes and Towels Wipes for General Applications
These economical, non-woven, polyester cellulose wipes are perfect for a variety of everyday and specialized tasks, especially in support work areas.
  • Creped surface of the wipe makes it more effective in picking up particulates and contaminants
  • Polyester cellulose blend delivers strength and absorbency
  • Versatile wiping and spill control
  • Highly absorbent, absorbs five times (5X) its own weight
Highly absorbent wipes are for less critical applications.
Applications include: Cleaning in support areas, parts and equipment manufacturing and maintenance, critical painting and sealing, routine maintenance and repair, surface and parts cleaning.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty