Would you like to log in again?
ACE1 Colorimetric Activity Assay Kit, BioVision
Supplier: BioVision
AB273308-100TEST
76375-830EA 909.88 USD InStock
76375-830
ACE1 Colorimetric Activity Assay Kit, BioVision
Assays Enzyme Assays
Able to detect ACE1 activity in a variety of samples.
  • Simple
  • Rapid
  • Sensitive method
Angiotensin-I-converting enzyme (ACE1) is a peptidyl dipeptidase that catalyzes the conversion of decapeptide angiotensin I to octapeptide angiotensin II, by removing a carboxy-terminal dipeptide. ACE is a key part of the renin angiotensin system (RAS) that regulates blood pressure. Increased ACE activity leads to an increase in the angiotensin II (Ang II) levels.
This may increase the risk of developing neurodegenerative diseases including Parkinson, Alzheimer, Huntington, Multiple Sclerosis etc. Elevated levels of Ang II mediate cardiac remodeling (cardiac hypertrophy and fibrosis), which is a hallmark in heart failure. Previous researchers have shown that ACE activity is a better predictor of pathologies associated with Type 2 diabetes than ACE levels in serum samples.
BioVision’s ACE1 Activity Assay Kit can be used to detect its activity in biological samples. This kit utilizes the ability of an active ACE1 to hydrolyze a synthetic substrate, which results in the decrease in optical density at 345 nm. The assay kit provides a rapid, simple and sensitive method to detect ACE1 activity as low as 40 mU in a variety of samples.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty