Would you like to log in again?
Glucosylceramidase Activity Assay Kit (Fluorometric), BioVision
Supplier: BioVision
AB273339-100TEST
76375-832EA 968.9 USD InStock
76375-832
Glucosylceramidase Activity Assay Kit (Fluorometric), BioVision
Assays Protein Assays
Able to monitor Glucosylceramidase activity.
  • Simple and rapid
Lysosomal glucosylceramidase beta also known as acid beta-glucosidase (E.C. 3.2.1.45) is a lysosomal membrane protein that cleaves the beta-glucosidic linkage of glycosylceramide (GC). The enzyme plays a central role in the degradation of complex lipids and the turnover of cellular membranes.
Through the production of ceramides, it participates in the PKC-activated salvage pathway of ceramide formation. Glucosylceramidase also plays a role in cholesterol metabolism. It may catalyze the glucosylation of cholesterol through a transglucosylation reaction that transfers glucose from glucosylceramide to cholesterol. Deficiency of Glucosylceramidase results in the accumulation of GC in lysosomes. This leads to Gaucher disease, an inherited disorder that affects many organs and tissues.
BioVision’s Glucosylceramidase Activity Assay Kit provides a simple, rapid way to monitor Glucosylceramidase activity in a wide variety of biological samples. In this kit, Glucosylceramidase cleaves a specific synthetic substrate and releases a fluorophore, which can be easily quantified (Ex/Em=360/445 nm). The assay is specific, sensitive and can detect as low as 0.2 µU of Glucosylceramidase activity in variety of samples.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty