Would you like to log in again?
EZClick™ Stearoylated Protein Assay Kit (FACS/Microscopy), Green Fluorescence, BioVision
Supplier: BioVision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
AB273283-100TEST
76375-840EA 786.88 USD InStock
76375-840
EZClick™ Stearoylated Protein Assay Kit (FACS/Microscopy), Green Fluorescence, BioVision
Assays Protein Assays
Good for labeling and detection of Stearoylated proteins.
  • Highly specific
  • Simple procedure
Stearoylation is one of the numerous types of post-translational modification in proteins. Stearoylated proteins are involved in regulation of cellular processes such as apoptosis, growth and metabolism. In this modification, Stearic acid, a saturated fatty acid, is covalently bound to a protein through the action of long-chain-fatty-acid-[acyl-carrier-protein] ligase. In the presence of ATP, this enzyme transfers a Stearate moiety to the target protein forming a carbon-sulfur bond (C=S).
Recent studies showed a possible correlation between mitochondrial dysfunction with some neurodegenerative diseases, cancer and aging. Furthermore, it is known that stearoylated Transferrin Receptor (TfR1) inhibits JNK activation, reduces mitofusin ubiquitination, thus restoring mitochondrial function. Studying of stearoylation of proteins can shed a light for disease etiology and prevention.
BioVision offers EZClick™ Stearoylated Protein Assay Kit, a highly specific, simple and robust method for labeling and detection of Stearoylated proteins. The kit uses Alkynyl Stearic Acid that is fed directly into the cells and gets incorporated into proteins during or post translation. This modification can be followed by click reaction with an azide-containing dye. The assay kit offers a powerful method for imaging localization, trafficking, and dynamics of Stearoylated proteins, or detection by FACS for quantitative studies.
Labeled Stearoylated Proteins can be directly detected in 1D or 2D gels using the appropriate excitation sources, or enriched by immunoprecipitation with biotin-azide or antibodies prior to proteomic analysis. We provide sufficient materials for 100 assays in a 96-well plate format.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty