Would you like to log in again?
EZClick™ Myristoylated Protein Assay Kit (FACS/Microscopy), Green Fluorescence, BioVision
Supplier: BioVision
AB273290-100TEST
76375-844EA 787.12 USD InStock
76375-844
EZClick™ Myristoylated Protein Assay Kit (FACS/Microscopy), Green Fluorescence, BioVision
Assays Protein Assays
Good for labeling and detection of myristoylated proteins
  • Simple procedure
  • Highly specific
Myristoylation is a co- and post-translational modification of proteins. Myristoylated proteins are involved in cellular trafficking, some cellular functions and oncogenesis. In this modification, Myristic acid, a saturated fatty acid is covalently bond to a protein through the action of N-myristoyl transferase (NMT). NMT transfers the myristate moiety from Myristoyl-CoA to a N-terminal glycine of a target protein. NMT is associated with carcinogenesis, the upregulation of its expression and activity is observed in colorectal cancer, gall-bladder cancer and brain tumors.
BioVision offers EZClick™ Myristoylated Protein Assay Kit, a highly specific, simple and robust method for labeling and detection of myristoylated proteins. We use Alkynyl Myristic Acid that is fed directly into the cells and gets incorporated into proteins during or post translation, followed by click reaction with and azide-containing dye.
This system offers a powerful method for imaging localization, trafficking, and dynamics of myristoylated proteins, or detection by FACS for quantitative studies. Labeled myristoylated proteins can be directly detected in 1D or 2D gels using the appropriate excitation sources, or enriched by immunoprecipitation with biotin-azide or antibodies prior to proteomic analysis. We provide sufficient materials for 100 assays in a 96-well plate format.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty