Would you like to log in again?
RNase Activity Detection/Quantification Assay Kit (Fluorometric), BioVision
Supplier: BioVision
AB273299-100TEST
76375-846EA 774.59 USD InStock
76375-846
RNase Activity Detection/Quantification Assay Kit (Fluorometric), BioVision
Assays Nucleic Acid Assays
Good for measuring RNase activity in buffers, reagents, and other components.
  • Highly sensitive
  • High throughput compatible
  • Specific
  • The limit of quantification is 1.2 pg RNase/well
  • The limit of detection is 0.4 pg RNase/well
Many molecular biology applications require RNase-free environment due to the degradation of RNA samples by RNase. Obtaining full length, high quality RNA samples is essential, yet challenging in genomics settings. BioVision’s RNAse Activity Detection Assay Kit enables researchers to measure RNase activity in buffers, reagents, and other components as well as quantitatively evaluate RNAse activity of recombinant enzymes in real time.
The assay uses a highly sensitive, specific probe that releases a fluorescent product in the presence of active RNAse. The limit of detection is 0.4 pg RNase/well and limit of quantification is 1.2 pg RNase/well.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty