Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

KIMBLE® Distillation Head with Condenser, DWK Life Sciences

Supplier: DWK Life Sciences
KIMBLE®
514000-2440
KT514000-2440EA 474.02 USD InStock
KT514000-2440
KIMBLE® Distillation Head with Condenser, DWK Life Sciences
Distillation Components Distillation Heads
Distillation head with a condenser and full length standard taper joints.

  • Simple take-off unit with a condenser sealed to the adapter at a 75° angle
  • With bottom and side standard taper inner drip joints
  • Standard taper 10/30 outer joint on top accepts a 75 mm immersion thermometer
  • Manufactured from 33 expansion, low extractable borosilicate glass

Hose connections are designed to fit ³/₈" ID flexible tubing.

Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty