Would you like to log in again?
Illinois Sternal/Iliac Bone Marrow Aspiration Needles, BD
Supplier: BD Medical
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
DIN1515X DIN1518X
PHDIN1515XCS 574.81 USD InStock
PHDIN1515X PHDIN1518X
Illinois Sternal/Iliac Bone Marrow Aspiration Needles, BD
Syringe Needles
The disposable intraosseous infusion needle from CareFusion provides the ability to gain vascular access for delivering fluids and drugs to critically ill or injured children
  • Sharp lancet point to facilitate entry into the bone
  • Luer slip/luer lock connection for a secure fit to any syringe
  • Adjustable depth guard to help control depth in needle insertion
  • Two sizes to meet users’ preference
Intraosseous infusion (IO) is recommended only in life-threateningemergencies where vascular access is critical and conventional forms of access are not available or are unobtainable.
Packaging: Needles are packaged sterile. Ten needles per case.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty