Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
BBL™ Taxo™ Anaerobe Differentiation Discs Set, BD Diagnostics
Supplier: BD Diagnostics
BBL™
231651 231651
BB4331651PK 166.37 USD InStock
BB4331651 BB433165
BBL™ Taxo™ Anaerobe Differentiation Discs Set, BD Diagnostics
Microbiology Test Systems Antibiotic Tests
These discs are recommended for use in the presumptive identification of gram-negative anaerobic bacilli based on differences in susceptibility to antimicrobial agents.
These discs are used for the differentiation of gram-negative anaerobic bacilli on the basis of in vitro testing of pure cultures against selected antimicrobial agents using the agar diffusion method. Growth of the microorganisms may be inhibited by the antimicrobial agent diffusing out from the disc, resulting in a zone of inhibition around the disc.
These ¹/₄" discs are made from high quality absorbent paper impregnated with accurately determined amounts of antimicrobial agents. They are furnished in cartridges containing 50 discs each. The last disc in each cartridge is marked 'X' and contains the drug as coded.
Store containers of discs at –20 to 8 °C.
Delivery information: Set components - one cartridge of 50 discs of each of the antimicrobial agents like BBL™ taxo™ Kanamycin, 1 mg, BBL™ taxo™ Rifampin, 15 µg, Sensi-Disc™ penicillin, 2 units, Sensi-Disc™ vancomycin, 5 µg and Sensi-Disc™ colistin, 10 µg.
Caution: The taxo Kanamycin 1 mg and taxo Rifampin 15 µg discs are not for use in susceptibility testing for therapeutic purposes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty