Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

BBL™ Trypticase™ Soy Agar, BD Life Sciences

Supplier: BD Life Sciences
BBL™
292385 215194 292385
95037-750PK 519.07 USD InStock
95037-750 CA-90004-268 90004-268
BBL™ Trypticase™ Soy Agar, BD Life Sciences
Media for Microbiology Prepared Media for Microbiology
Trypticase™ soy agar mainly for isolation and cultivation of non-fastidious and fastidious microorganisms.

  • Liquid media
  • Color - light beige

Store at 2 to 8°C.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty