Would you like to log in again?
Humidification Chamber for Single Use, BD
Supplier: BD Medical
MR250
PHMR250CS 2902.65 USD InStock
PHMR250
Humidification Chamber for Single Use, BD
Clinical Diagnostic Supplies
Humidification chamber for single use. Designed for adult patients and allows for manual filling.
  • Easy to use
  • Slide onto all humidifier heater bases
  • For adults
  • Offers high-flow performance
Packaging: 40 per case.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty