Would you like to log in again?
DURAN® Laboratory Bottles, Pressure plus+, DWK Life Sciences
Supplier: DWK Life Sciences
DURAN®
218105403 218102406 218165409 1092235 1094367 1094368 1092234
76263-730CS 321.88 USD InStock
76263-730 76263-728 76264-176 76263-732 75995-260 76264-180 76263-734
DURAN® Laboratory Bottles, Pressure plus+, DWK Life Sciences
Bottles Glass Bottles
With easy-to-read scale and large labelling field for easy marking, in fired-on, highly durable ceramic.
  • DIN thread, GL 45
  • Retrace code
  • Autoclavable
When pressure loaded the following apply: thermal shock resistance 30 K and maximum usage temperature +140 °C. Blue scale for visual differentiation from the standard laboratory bottle. UV protection up to approx. 500 nm wavelength. Unchanged DURAN® properties within the bottle, as colouration is only on the outer surface.
Typical applications: safe working under pressure or vacuum, sampling under pressure, storage of gas generating media.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty