Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

n-Hexane

Supplier: Pfaltz & Bauer
Synonyms: HexanenHexaneHexyl hydride

H05280-500G
101186-810EA 112.46 USD InStock
101186-810
n-Hexane
n-Hexane


Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 69 °C
Melting Pt: –95 °C
Density: 0.659 g/ml
Flash Pt: –26 °C
MDL Number: MFCD00009520
CAS Number: 110-54-3
UN: 1208
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/70/18/737018.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty