Would you like to log in again?
Natural Bristles Beaker Brush
Supplier: United Scientific Supplies
BBR016
470320-988EA 12.45 USD InStock
470320-988
Natural Bristles Beaker Brush
Brushes
Good quality natural black bristles, mounted in twisted heavy gauge galvanized wire with radial top.
  • Natural bristles
  • Radial top
This natural bristle brush reaches into all the corners to give you a clean beaker. The radial tops ensures you do not scratch the glass.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty