Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
LiChroCART® Purospher® HPLC Columns, STAR RP-8e, MilliporeSigma
Supplier: MilliporeSigma
LiChroCART® Purospher®
1.50171.7194 1.50171.7194
10144-056EA 706.62 USD InStock
10144-056 CA1.50171.7194
LiChroCART® Purospher® HPLC Columns, STAR RP-8e, MilliporeSigma
Chromatography Columns
Stainless steel LiChroCART® cartridges are available with various types of Purospher® sorbents.
These LiChroCART® cartridges require part number 14229-736 manu-CART® cartridge column holder, which can be used to hold one cartridge column with or without a 4×4 mm guard column. LiChroCART® columns 250×10 mm require part number 48219-720 manu-CART® 10. The short LiChroCART® columns can be ordered as a set including the corresponding cartridge holder and one cartridge, or as a pack of 3 cartridges without cartridge holder. The separate part numbers for the cartridge are as follows: 48219-758 LiChroCART® cartridge holder for 30 mm cartridge and 48219-756 LiChroCART® cartridge holder for 55 mm cartridge.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty