Would you like to log in again?
Flame Resistant Insulated Bomber Jackets, MCR Safety
Supplier: MCR Safety
B3NM B3NS
76341-944EA 483.78 USD InStock
76341-944 76341-868
Flame Resistant Insulated Bomber Jackets, MCR Safety
Workwear
Whether working in the rugged oil and gas industry or spending the day working near electrical hazards, stay safe and comfortable in FR gear made with Max Comfort™ fabrics.
  • Max Comfort™ 7.5 oz., 88% cotton/12% nylon outer shell
  • 11.5 oz. quilted lining, 100% FR treated cotton and non-woven modacrylic batting
  • Water repellent
  • Sewn with FR aramid thread
  • Double needle stitching and tacking for reinforcement
The B3N insulated bomber jacket features a comfortable 88% cotton/12% nylon material construction with a modacrylic quilted lining for warmth and protection.
Certifications: Max Comfort™ Fabric NFPA 2112 UL Certified; All Trims are NFPA 2112 UL Certified; NFPA 70E Hazard/Risk Category 3 (HRC3), Arc Flash PPE Category 3 (CAT3); ASTM F1506 Electric Arc Protection Standard; ATPV ASTM 1959 35 cal/cm², HRC3 / CAT3; Bomber Jacket is UL & NFPA 2112 Certified. Max Comfort™ material meets the NFPA 2112-2018 Standard for Flame Resistant Garments and NFPA 70E Standard for Electrical Safety in the workplace.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty