Would you like to log in again?
Flame Resistant Insulated Parkas, MCR Safety
Supplier: MCR Safety
PK3NX5 PK3NL PK3GL PK3NX4T PK3GX4T PK3GX4 PK3GX5T PK3GX3T PK3NLT PK3NS PK3GX2T PK3GLT PK3GX5 PK3NXLT PK3NM PK3NX3T PK3GS PK3NX3 PK3NX2T PK3NX2 PK3GM PK3NX5T PK3NX4
76342-308EA 507.36 USD InStock
76342-308 76342-070 76342-196 76342-372 76342-370 76342-260 76342-194 76342-062 76342-166 76342-364 76342-266 76342-016 76342-302 76342-236 76342-358 76342-402 76342-314 76342-028 76342-410 76342-422 76342-106 76342-436 76342-118
Flame Resistant Insulated Parkas, MCR Safety
Workwear
Whether working in the rugged oil and gas industry or spending the day working near electrical hazards, stay safe and comfortable in FR gear made with Max Comfort™ fabrics.
  • Max Comfort™ 7.5 oz., 88% cotton/12% nylon outer shell
  • 11.5 oz. quilted lining, 100% FR treated cotton and non-woven modacrylic batting
  • Water repellent
  • Two patch pockets on chest with hidden snap closures
  • Inside chest pocket
  • Pencil stall on left sleeve
The insulated parka is manufactured from a comfortable 88% cotton/12% nylon twill material with a modacrylic quilted lining for warmth and protection.
Certifications: Max Comfort™ Fabric NFPA 2112 UL certified. All trims are NFPA 2112 UL certified. NFPA 70E hazard/risk Cat HRC3. ASTM F1506 electric arc protection standard. ATPV ASTM 1959 35 cal/cm², HRC 3. Parka is UL and NFPA 2112 certified. Max Comfort™ material meets the NFPA 2112-2018 standard for flame resistant garments and NFPA 70E standard for electrical safety in the workplace.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty