Would you like to log in again?
Flame Resistant Hooded Sweatshirts, MCR Safety
Supplier: MCR Safety
Max Comfort™
SS1NX4 SS1NX3 SS1NS SS1NXL SS1NM SS1NX2 SS1NL SS1NX5
76341-410EA 353.15 USD InStock
76341-410 76341-350 76341-548 76341-516 76341-208 76341-510 76341-412 76341-258
Flame Resistant Hooded Sweatshirts, MCR Safety
Workwear
Whether working in the rugged oil and gas industry or spending the day working near electrical hazards, stay safe and comfortable in FR gear made with Max Comfort™ fabrics.
  • Max Comfort™ material
  • 100% FR cotton interlock fleece shell for maximum comfort
  • Sewn with flame resistant thread
The Max Comfort™ material meets the NFPA 2112-2018 standard for flame resistant garments and NFPA 70E standard for electrical safety in the workplace.
Certifications: NFPA 2112 UL certified; ASTM F1506 electric arc protection; ASTM 1959; ATPV 18 cal/cm².
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty