Would you like to log in again?
Memphis Welding Leather Welding Sleeves with Snap, MCR Safety
Supplier: MCR Safety
38418MW
76340-866PR 60.55 USD InStock
76340-866
Memphis Welding Leather Welding Sleeves with Snap, MCR Safety
Workwear
Heavy select gray side-split leather provides outstanding protection from heat, sparks, slag, and cuts.
  • Select gray split cow leather
  • Double lock stitched, sewn with Kevlar®
  • Sleeve with snaps
Sleeves are ideally suited for use in welding, construction, Heliarc, iron work, metal fabrication and Mig/Tig applications.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty