Would you like to log in again?
Dominator Series PVC Aprons, MCR Safety
Supplier: MCR Safety
388R5
76342-604DZ 188.28 USD InStock
76342-604
Dominator Series PVC Aprons, MCR Safety
Workwear
This heavy-duty, thick PVC apron provides outstanding durability.
  • Thick PVC material construction
  • 0.42 mm thickness
  • Great for splash protection, abrasion and puncture protection
  • Durable plastic reinforced grommets, with tie strings included
  • Raw edges
This apron measures 35×48", has raw edges, and is aqua green in color. This garment is ideally suited for use in agriculture, assembly line production, bottling, cafeteria, dairy, janitorial, veterinary, beef processing, fisheries, pork processing, and poultry processing applications.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty