Would you like to log in again?
Memphis Welding 45" Welding Jackets, MCR Safety
Supplier: MCR Safety
Memphis Welding
39045XL 39045L
76340-990EA 76.12 USD InStock
76340-990 76341-042
Memphis Welding 45" Welding Jackets, MCR Safety
Workwear
These jackets provide economical protection from exposure to molten splash and sparks.
  • USA treated cotton fabric
  • Anodized button snap closures
All limited flammability cotton apparel is made of 9 oz. green cotton. The 100% cotton material is cool, comfortable, and launderable. Jackets will maintain limited flammability up to 50 home washings or 25 commercial washings (up to 165°).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty