Would you like to log in again?
BioClean Dual™ Disposable Cleanroom Shoe Cover, PP/CPE, Nitritex, Ansell
Supplier: Ansell
S-BDOS
75982-218CS 637.38 USD InStock
75982-218
BioClean Dual™ Disposable Cleanroom Shoe Cover, PP/CPE, Nitritex, Ansell
Disposable Garments
Disposable shoe covers made from BioClean Dual™, a low linting spunbonded non-woven polypropylene fabric with embossed cast polyethylene sole for a secure footing.
  • Durable
  • Heavy-duty, slip-resistant sole
  • Latex-free elastic
Suitable for use in ISO Class 4 environment.
Certifications: Manufactured in a facility holding ISO 9001:2008 certification and processed in an NEBB certified ISO Class 4 environment.
Packaging: One pair per sealed inner PE bag, 15 inner bags per sealed outer bag, 10 outer bags per lined carton (150 pairs).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty