Would you like to log in again?
O-Ring Vacuum Connector with Hose Connection, DWK Life Sciences
Supplier: DWK Life Sciences
183100-1502
KT183100-1502EA 117.85 USD InStock
KT183100-1502
O-Ring Vacuum Connector with Hose Connection, DWK Life Sciences
Fittings and Connectors
This O-ring connector of size 5 mm ID for use with HI-VAC® manifolds and adapters.
  • For applications utilizing tubing
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Delivery information: Supplied with one size 116 FKM O-ring.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty