Would you like to log in again?
AIRLESS-WARE® Filling Adapter, DWK Life Sciences
Supplier: DWK Life Sciences
211230-2440
KT211230-2440EA 151.45 USD InStock
KT211230-2440
AIRLESS-WARE® Filling Adapter, DWK Life Sciences
Adapters Adapters, Joints with Tube Connectors
This adapter is designed for use with a solvent repurifier to facilitate product removal from the storage area.
  • Provided clearance from the boiling flask and attaches to a solvent storage flask
  • Hose connection allows access to vacuum source or inert gas
Manufactured from 33 expansion, low extractable borosilicate glass.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty