Would you like to log in again?
KIMBLE® KONTES® Condenser, Micro, Allihn, DWK LIFE SCIENCES
Supplier: DWK Life Sciences
283250-0000
KT283250-0000EA 199.36 USD InStock
KT283250-0000
KIMBLE® KONTES® Condenser, Micro, Allihn, DWK LIFE SCIENCES
Condensers
Allihn style micro condenser is used in refluxing operations such as Soxhlet extractions. Component of Soxhlet extraction apparatus KT292010-0000.
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty