Would you like to log in again?
WHEATON® Pasteur Pipette Adapter, DWK Life Sciences
Supplier: DWK Life Sciences
W820125
75800-372CS 69.69 USD InStock
75800-372
WHEATON® Pasteur Pipette Adapter, DWK Life Sciences
Pipettes Micropipettes
PVDF adapter fits pipette nozzle to accommodate standard glass Pasteur pipettes for model 832 and 835.
The adapter has two FPM O-rings guaranteeing proper holding and tightness of standard glass 2 ml Pasteur pipettes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty