Would you like to log in again?
Liebig Condenser, DWK Life Sciences
Supplier: DWK Life Sciences
282210-0000
KT282210-0000EA 195.61 USD InStock
KT282210-0000
Liebig Condenser, DWK Life Sciences
Condensers
Liebig condenser is mainly designed to eliminate the use of bulbs where holdup is likely to occur.
Lower inner drip joint is designed to extend into 287800-0000 distillation receiver.
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Conforms to NSN 6640-01-346-9222.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty