Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DURAN® Pouring Rings, DWK Life Sciences

Supplier: DWK Life Sciences
DURAN®
292441909 292442802
76264-256PK 41.85 USD InStock
76264-256 76264-254
DURAN® Pouring Rings, DWK Life Sciences
Closures Bottle Closures
Red melamine resin rings for pouring accuracy.

  • Spill prevention when pouring from laboratory bottles
  • ETFE
  • Temp. stability range: –100 to 180 °C
  • Autoclavable at 121 °C

Permits clean, drop-free use.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty