Would you like to log in again?
KIMBLE® O-Ring Glass Connector Cap, DWK Life Sciences
Supplier: DWK Life Sciences
KIMBLE®
851810-0015
10135-814CS 47.88 USD InStock
10135-814
KIMBLE® O-Ring Glass Connector Cap, DWK Life Sciences
Fittings and Connectors
O-ring connector to seal flasks or for use with HI-VAC® manifold.
  • Supplied with one size 116 FKM O-ring
Manufactured from 33 expansion, low extractable borosilicate glass.
Certifications: Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty