Would you like to log in again?
DURAN® YOUTILITY, Bottle Tags, DWK Life Sciences
Supplier: DWK Life Sciences
DURAN®
292432834 292432867 292432883 292432818 292432875 292432842 292432826 292432859
76264-240PK 98.57 USD InStock
76264-240 76264-250 76264-244 76264-242 76264-248 76264-238 76264-246 76264-236
DURAN® YOUTILITY, Bottle Tags, DWK Life Sciences
Labels
These bottle tags are used for easy customization and unambiguous bottle identification.
  • Fits thread GL 45
  • Securely attaches around the neck of the bottle
  • Elastic for flexibility
  • Wide range of colors available
  • Autoclavable at 121 °C
The colorful silicone GL 45 bottle tags can be easily and securely attached around the neck of the YOUTILITY bottle. The Bottle Tags are available in several distinct colors for easy customization and unambiguous bottle identification. The elastic bottle tags will fit around any DIN GL 45 bottle neck, and can even be applied with the screw cap in place.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty