Would you like to log in again?
KIMBLE® Flex Shaft Adapters, DWK Life Sciences
Supplier: DWK Life Sciences
788020-0021
KT788020-0021EA 198.2 USD InStock
KT788020-0021
KIMBLE® Flex Shaft Adapters, DWK Life Sciences
Fittings and Connectors
This convenient stainless-steel universal joint with ⁵/₁₆" O.D. shaft and polyacetal adapter connects stirrer shafts to stirrer motors.
  • Fits shaft Ø: 8 to 10 mm
Flexible design makes the connection better even the alignment is not precise.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty