Would you like to log in again?
WHEATON® MicroLiter Shell Vial Kits, DWK Life Sciences
Supplier: DWK Life Sciences
96-2010N
30618-480PK 63.96 USD InStock
30618-480
WHEATON® MicroLiter Shell Vial Kits, DWK Life Sciences
Vials
MicroLiter offers products for both LC and GC applications.
  • Shell of 1 ml
  • Amber glass
This kit is conveniently packaged with a shell amber vial and a push-in cap.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty