Would you like to log in again?
KIMBLE® Filtration Domes, DWK Life Sciences
Supplier: DWK Life Sciences
KIMBLE®
953870-2000 953870-1000
KT953870-1000EA 1156.06 USD InStock
KT953870-1000 KT953870-2000
KIMBLE® Filtration Domes, DWK Life Sciences
Filters Bottle Top Filters
Filter domes are recommended for EPA-type sample digestions prior to atomic absorption spectroscopy.
  • Constructed of non-metallic materials
  • Filters quickly and simply, three times faster than the gravity method
  • Filtrate is collected directly in a beaker, eliminating transfers
  • Designed for direct filtration and for use where transfers should be avoided
  • Valve and hose connections are made of CTFE
  • O-rings are FKM
Filter domes are constructed of 33 expansion, low extractable borosilicate glass and polypropylene, virtually eliminating chances of trace metal contamination.
Several units may be connected in a series and vacuum applied from a single source. For rubber stopper size eight.
Certifications: Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty