Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ultra-Low Temperature Chambers, −40 to −80 °C, Farrar Scientific

Supplier: Farrar Scientific
ULCI-80C-W ULCI-80C
76377-658EA 164937.63 USD InStock
76377-658 76377-656
Ultra-Low Temperature Chambers, −40 to −80 °C, Farrar Scientific
Freezers Ultra Low Temperature Freezers
Forced air circulation ULT freezers specifically designed for pharmaceutical, biorepository/biobank.

  • Largest volume space
  • Forced-air circulation provides fastest temperature recovery from door opening
  • Fully redundant systems offer peace of mind
  • Small foot print
  • Low temperature deviation (±1 ˚C)
  • 288000 vial storage space

Designed to balance energy and have a dynamic refrigeration control.

Low energy consumption 0.33 kWh/cu.ft.

Chamber volume: 190 cu.ft./5380 L

Alarm system for high/low temperature, door ajar, power failure, remote alarm contacts.

Ordering information: Two year warranty.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty