Would you like to log in again?
Connecting Adapter, DWK Life Sciences
Supplier: DWK Life Sciences
747160-0000
KT747160-0000EA 31.81 USD InStock
KT747160-0000
Connecting Adapter, DWK Life Sciences
Adapters Adapters, Joints with Tube Connectors
Glassware is manufactured from 33 expansion, low extractable borosilicate glass.
  • Ideal for use to 200 °C
They can be assembled 'right side up' or 'upside down', providing great versatility.
Certifications: Conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty