Would you like to log in again?
KIMBLE® ULTRA-WARE® Standard Fittings Kit, DWK Life Sciences
Supplier: DWK Life Sciences
ULTRA-WARE®
953882-1000
KT953882-1000EA 782.86 USD InStock
KT953882-1000
KIMBLE® ULTRA-WARE® Standard Fittings Kit, DWK Life Sciences
Fittings and Connectors
This kit contains all of the tube fittings, adapters and solvent inlet filters required to plumb any ULTRA-WARE® cap to virtually all HPLC pumps.
Adapter, F 1/4-28 to F Luer (420407-2003), adapter, M 1/4-28 to M Luer (420407-2113), union, 5/16-24 to 1/4-28 (420818-3305), tubing nut, 1/4-28, for ¹/₈" OD (420821-0018), tubing nut, M6, for ¹/₈" OD (420821-0618), tubing nut, 5/16-24, for ³/₁₆" OD (420821-3519), ferrule, for ¹/₈" OD (420822-0018), ferrule, for ³/₁₆" OD (420822-1019), ferrule, for 1.8 mm OD (420822-2007), inlet filter, for ¹/₈" OD tubing (953916-0000), inlet filter, BOTB, for ¹/₈" OD tubing (953916-1002), adapter, 1/4-28 to 1/4" hose barb (953919-0014). Not recommended for use with THF or chlorinated hydrocarbons.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty